missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Follicle-stimulating hormone receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Follicle-stimulating hormone receptor, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6057-ec-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_000136.2 | |
Follicle-stimulating hormone receptor Protein | |
Research Use Only | |
FSHR | |
Recombinant Protein | |
E. coli | |
CHHRICHCSN RVFLCQESKV TEIPSDLPRN AIELRFVLTK LRVIQKGAFS GFGDLEKIEI SQNDVLEVIE ADVFSNLPKL HEIRIEKANN LLYINPEAFQ NLPNLQYLLI SNTGIKHLPD VHKIHSLQKV LLDIQDNINI HTIERNSFVG LSFESVILWL NKNGIQEIHN CAFNGTQLDE LNLSDNNNLE ELPNDVFHGA SGPVILDISR TRIHSLPSYG LENLKKLRAR STYNLKKLPT LEKLVALMEA SLTYPSHCCA FANWRRQISE LHPICNKSIL RQEVDYMTQA RGQRSSLAED NESSYSRGFD MTYTEFDYDL CNEVVDVTCS PKPDAFNPCE DIMGYNILR | |
Greater than 90% as determined by SDS-PAGE. |
2492 | |
1 mg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |