missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FLT3L / Flt3 ligand Protein
A DNA sequence encoding the Human FLT3L / Flt3 ligand, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10358-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001191431.1 | |
FLT3L / Flt3 ligand Protein | |
Research Use Only | |
FLT3LG | |
The ED50 as determined by a cell proliferation assay using human AML5 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.0, lyophilized |
2323 | |
10 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TQDCSFQHSP ISSDFAVKIR ELSDYLLQDY PVTVASNLQD EELCGGLWRL VLAQRWMERL KTVAGSKMQG LLERVNTEIH FVTKCAFQPP PSCLRFVQTN ISRLLQETSE QLVALKPWIT RQNFSRCLEL QCQPDSSTLP PPWSPRPLEA TAPTA | |
Greater than 97% as determined by SDS-PAGE and HPLC. |