missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroblast growth factor 23 Protein

A DNA sequence encoding the Human Fibroblast growth factor 23, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10385-ec-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983688

  • 313.00 CHF / 20 Mikrogramm


Verfügbar ab
02-11-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_065689.1
Fibroblast growth factor 23 Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
YPNASPLLGS SWGGLIHLYT ATARNSYHLQ IHKNGHVDGA PHQTIYSALM IRSEDAGFVV ITGVMSRRYL CMDFRGNIFG SHYFDPENCR FQHQTLENGY DVYHSPQYHF LVSLGRAKRA FLPGMNPPPY SQFLSRRNEI PLIHFNTPIP RRHTRSAEDD SERDPLNVLK PRARMTPAPA SCSQELPSAE DNSPMASDPL GVVRGGRVNT HAGGTGPEGC RPFAKFI
8074
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
FGF23
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ug/ml, corresponding to a specific activity of > 2.0 M 103 IU/mg in the presence of 0.3 ug/ml of rMuKlotho and 10 ug/ml of heparin.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation