missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF16 Protein
A cDNA sequence encoding the FGF16 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | FGF16 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biologische Aktivität | The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. |
Produkttyp | Recombinant Protein |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15964378
|
enQuireBio™
QP10601-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15954378
|
enQuireBio™
QP10601-25UG |
25 μg |
203.00 CHF
25 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15944378
|
enQuireBio™
QP10601-1MG |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
FGF16 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AEVGGVFASL DWDLHGFSSS LGNVPLADSP GFLNERLGQI EGKLQRGSPT DFAHLKGILR RRQLYCRTGF HLEIFPNGTV HGTRHDHSRF GILEFISLAV GLISIRGVDS GLYLGMNERG ELYGSKKLTR ECVFREQFEE NWYNTYASTL YKHSDSERQY YVALNKDGSP REGYRTKRHQ KFTHFLPRPV DPSKLPSMSR DLFHYR | |
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg. | |
Human | |
Untagged | |
FGF-16 0.2microm filtered solution containing 20mM Tris-HCl, 1M NaCl, pH 9.0, 0.2% Tween-20 and 10% Glycerol. |