missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGA Protein
A DNA sequence encoding the Homo sapiens (Human) FGA, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6034-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
FGA Protein | |
Research Use Only | |
FGA | |
Recombinant Protein | |
E. coli | |
GPRVVERHQS ACKDSDWPFC SGEDWNYKCP SGCRMKGLID EVNQDFTNRI NKLKNSLFEY QKNNKDSHSL TTNIMEILRG DFSSANNRDN TYNRVSEDLR SRIEVLKRKV IEKVTANNLL VARVTTEETP HLKARAIKWQ MRPEVKPIMK EHIAPREAML NLALSEVSTL LLWGSLPCPP RLS | |
Greater than 80% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |