missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FDCSP Protein
A cDNA sequence encoding the FDCSP was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11863-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
FDCSP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFPVSQDQ EREKRSISDS DELASGFFVF PYPYPFRPLP PIPFPRFPWF RRNFPIPIPE SAPTTPLPSE K | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FDCSP protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 0.15M NaCl, 50% glycerol and 1mM DTT. |