missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM19A2 Protein

A DNA sequence encoding the Human FAM19A2, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10244-ec-20ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983208

  • 313.00 CHF / 20 Mikrogramm


Verfügbar ab
28-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_848634.1
FAM19A2 Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ANHHKAHHVK TGTCEVVALH RCCNKNKIEE RSQTVKCSCF PGQVAGTTRA APSCVDASIV EQKWWCHMQP CLEGEECKVL PDRKGWSCSS GNKVKTTRVT H
338811
Greater than 95% as determined by SDS.­PAGE and HPLC.
Research Use Only
FAM19A2
The biological activity is determined by its ability to enhance neurite outgrowth of E16­E18 rat embryonic cortical neurons. rHuTAFA­2, immobilized at 6­24 ug/mL on a 96 well plate, is able to significantly enhance neurite outgrowth.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation