missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Erythropoietin / EPO Protein

A DNA sequence encoding the Human Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10347-ma-37500IU

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983578

  • 905.00 CHF / 37500 Einheiten


Verfügbar ab
26-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_000790.2
Erythropoietin / EPO Protein
37500 IU
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
Mammal
0.2um filtered sodium citrate buffer (1 liter of ddH2O containing 5.9 g of sodium citrate, 5.8 g of sodium chloride and 0.06 g of citric acid), lyophilized
2056
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
EPO
The ED50 determined by a cell proliferation assay using human megakaryoblastic leukemia cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
FC
Videos
Dokumentation