missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epigen Protein

A DNA sequence encoding the Human Epigen, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10396-ec-25ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983718

  • 313.00 CHF / 25 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001257918.1
Epigen Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
AVTVTPPITA QQADNIEGPI ALKFSHLCLE DHNSYCINGA CAFHHELEKA ICRCFTGYTG ERCEHLTLTS YA
255324
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
EPGN
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 300 ng/ml, corresponding to a specific activity of > 3.3 M 103 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation