missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENO3 / beta-enolase Protein
A DNA sequence encoding the Homo sapiens (Human) ENO3 / beta-enolase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP1054-ec-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001180432.1 | |
ENO3 / beta-enolase Protein | |
Research Use Only | |
ENO3 | |
Recombinant Protein | |
E. coli | |
FAREILDSRG NPTVEVDLHT AKGRFRAAVP SGASTGIYEA LELRDGDKGR YLGKGVLKAV ENINNTLGPA LLQKKLSVVD QEKVDKFMIE LDGTENKSKF GANAILGVSL AVCKAGAAEK GVPLYRHIAD LAGNPDLILP VPAFNVINGG SHAGNKLAMQ EFMILPVGAS SFKEAMRIGA EVYHHLKGVI KAKYGKDATN VGDEGGFAPN ILENNEALEL LKTAIQAAGY PDKVVIGMDV AASEFYRNGK YDLDFKSPDD PARHITGEKL GELYKSFIKN YPVVSIEDPF DQDDWATWTS FLSGVNIQIV GDDLTVTNPK RIAQAVEKKA CNCLLLKVNQ IGSVTESIQA CKLAQSNGWG VMVSHRSGET EDTFIADLVV GLCTGQIKTG APCRSERLAK YNQLMRIEEA LGDKAIFAGR KFRNPK | |
Greater than 90% as determined by SDS-PAGE. |
2027 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |