Learn More
enQuireBio™ Recombinant Human CXCL9 / MIG / C-X-C motif chemokine 9 Protein
Recombinant Protein
Marke:  enQuireBio™ QP1020-5 ug
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15983068
  • 98.65 CHF / 5 Mikrogramm
Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen
Spezifikation
Spezifikation
NP_002407.1
CXCL9 / MIG / C-X-C motif chemokine 9 Protein
5 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
TPVVRKGRCS CISTNQGTIH LQSLKDLKQF APSPSCEKIE IIATLKNGVQ TCLNPDSADV KELIKKWEKQ VSQKKKQKNG KKHQKKKVLK VRKSQRSRQK KTT
4283
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CXCL9
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA