missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL26 / Eotaxin 3 Protein
A DNA sequence encoding the Human CCL26 / Eotaxin 3, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10241-ec-250ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_006063.1 | |
CCL26 / Eotaxin 3 Protein | |
Research Use Only | |
CCL26 | |
The biological activity determined by a chemotaxis bioassay using human CCR3 transfected HEK293 cells is in a concentration range of 0.5- 2.0 ug/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
10344 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
TRGSDISKTC CFQYSHKPLP WTWVRSYEFT SNSCSQRAVI FTTKRGKKVC THPRKKWVQK YISLLKTPKQ L | |
Greater than 97% as determined by SDS-PAGE and HPLC. |