missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL21 / 6Ckine Protein
A DNA sequence encoding the Human CCL21 / 6Ckine, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10236-ec-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_002980.1 | |
CCL21 / 6Ckine Protein | |
Research Use Only | |
CCL21 | |
The biological activity determined by a chemotaxis bioassay using human lymphocytes is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6366 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SDGGAQDCCL KYSQRKIPAK VVRSYRKQEP SLGCSIPAIL FLPRKRSQAE LCADPKELWV QQLMQHLDKT PSPQKPAQGC RKDRGASKTG KKGKGSKGCK RTERSQTPKG P | |
Greater than 97% as determined by SDS-PAGE and HPLC. |