missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL13 / MCP-4 Protein
A DNA sequence encoding the Human CCL13 / MCP-4, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10233-ec-20ug
Weitere Details : Gewicht : 0.01000kg
Specifications
NP_005399.1 | |
CCL13 / MCP-4 Protein | |
Research Use Only | |
CCL13 | |
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6357 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
QPDALNVPST CCFTFSSKKI SLQRLKSYVI TTSRCPQKAV IFRTKLGKEI CADPKEKWVQ NYMKHLGRKA HTLKT | |
Greater than 96% as determined by SDS-PAGE and HPLC. |