missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human C-X-C motif chemokine 6 Protein

A DNA sequence encoding the Human C-X-C motif chemokine 6, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10211-ec-250ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983098

  • 2310.00 CHF / 250 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_002984.1
C-X-C motif chemokine 6 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VLTELRCTCL RVTLRVNPKT IGKLQVFPAG PQCSKVEVVA SLKNGKQVCL DPEAPFLKKV IQKILDSGNK KN
6372
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL6
The biological activity determined by a chemotaxis bioassay using human neutrophils is in a concentration range of 10-50 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Dokumentation