missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP10 Protein
A cDNA sequence encoding the BMP10 was constructed and used to recombinantly synthesize the protein.
119.00 CHF - 700.00 CHF
Spezifikation
Gen-ID (Entrez) | 27302 |
---|---|
Name | BMP10 Protein |
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gensymbol | BMP10 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15974168
|
enQuireBio™
QP10536-1UG |
1 μg |
119.00 CHF
1 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15984168
|
enQuireBio™
QP10536-5UG |
5 μg |
432.00 CHF
5 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15964168
|
enQuireBio™
QP10536-10UG |
10 μg |
700.00 CHF
10 Mikrogramm |
Verfügbar ab: 24-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
27302 | |
Research Use Only | |
BMP10 | |
Recombinant Protein | |
Human (HEK293) | |
NAKGNYCKRT PLYIDFKEIG WDSWIIAPPG YEAYECRGVC NYPLAEHLTP TKHAIIQALVHLKNSQKASK ACCVPTKLEP ISILYLDKGV VTYKFKYEGM AVSECGCR | |
Greater than 95.0% as determined by analysis by SDS-PAGE and HPLC analysis. |
BMP10 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The specific activity was determined by its ability to induce alkaline phosphatase production in the ATDC-5 cell line and is typically 4.0-6.0 ng/ml ED50. | |
Human | |
Untagged | |
The BMP10 protein was lyophilized from 10mM Sodium Citrate, pH 2.7 and 75mM NaCl . |