missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Beta-defensin 104 Protein

A DNA sequence encoding the Human Beta-defensin 104, was expressed in the hosts and tags indicated.

Marke:  enQuireBio™ QP10432-ec-5ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15983838

  • 123.00 CHF / 5 Mikrogramm


Verfügbar ab
30-10-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

NP_001035792.1
Beta-defensin 104 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
EFELDRICGY GTARCRKKCR SQEYRIGRCP NTYACCLRKW DESLLNRTKP
107984148
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
DEFB104A
The biological activity determined by a chemotaxis bioassay using human monocytes is in a concentration range of 0.1-100.0 ng/ml.
Human
Untagged
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized
Videos
Dokumentation