missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AUH Protein
A cDNA sequence encoding the AUH was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP11111-5ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
549 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSSEMKTEDE LRVRHLEEEN RGIVVLGINR AYGKNSLSKN LIKMLSKAVD ALKSDKKVRT IIIRSEVPGI FCAGADLKER AKMSSSEVGP FVSKIRAVIN DIANLPVPTI AAIDGLALGG GLELALACDI RVAASSAKMG LVETKLAIIP GGGGTQRLPR AIGMSLAKEL IFSARVLDGK EAKAVGLISH VLEQNQEGDA AYRKALDLAR EFLPQGPVAM RVAKLAINQG MEVDLVTGLA IEEACYAQTI PTKDRLEGLL AFKEKRPPRY KGE | |
Greater than 95.0% as determined by SDS-PAGE. |
AUH Protein | |
Research Use Only | |
AUH | |
Human | |
His | |
The AUH solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT. |