missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ASMT Protein
A cDNA sequence encoding the ASMT was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 5210.00 CHF
Spezifikation
Name | ASMT Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15986098
|
enQuireBio™
QP11087-5UG |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15976098
|
enQuireBio™
QP11087-20UG |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15966098
|
enQuireBio™
QP11087-1MG |
1 mg |
5210.00 CHF
1 Milligramm |
Verfügbar ab: 27-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
ASMT Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ASMT solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) , 1M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSEDQAYR LLNDYANGFM VSQVLFAACE LGVFDLLAEA PGPLDVAAVA AGVRASAHGT ELLLDICVSL KLLKVETRGG KAFYRNTELS SDYLTTVSPT SQCSMLKYMG RTSYRCWGHL ADAVREGRNQ YLETFGVPAE ELFTAIYRSE GERLQFMQAL QEVWSVNGRS VLTAFDLSVF PLMCDLGGDF FKDPLPEADL YILARVLHDW ADGKCSHLLE RIYHTCKPGG GILVIESLLD EDRRGPLLTQ LYSLNMLVQT EGQERTPTHY HMLLSSAGFR DFQFKKTGAI YDAILARK | |
Greater than 90.0% as determined by SDS-PAGE. |