missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AP1S2 Protein

A cDNA sequence encoding the AP1S2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP11039-2ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965948

Menge Preis
1 128.22 CHF / 2 Mikrogramm
10 117.77 CHF / 2 Mikrogramm
Voraussichtliches Versanddatum: 10-11-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

AP1S2 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
AP1S2 protein solution (0.5 mg/1ml) containing 20mM Tris-HCl buffer (pH 8.0), 40% glycerol, 0.1M NaCl and 2mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MQFMLLFSRQ GKLRLQKWYV PLSDKEKKKI TRELVQTVLA RKPKMCSFLE WRDLKIVYKR YASLYFCCAI EDQDNELITL EIIHRYVELL DKYFGSVCEL DIIFNFEKAY FILDEFLLGG EVQETSKKNV LKAIEQADLL QEEAETPRSV LEEIGLT
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate