missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ang K1-3 Protein

A cDNA sequence encoding the Ang K1-3 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10520-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15984118

  • 203.00 CHF / 100 Mikrogramm
Verfügbar ab: 29-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

Ang K1-3 Protein
Research Use Only
The activity is assayed on anti-proliferation and anti-migration of endothelial cells in vitro and anti-angiogenesis in vivo. The specific activity of anti-migration of endothelial cells in vitro is 55,000 Units/mg.
Human
Untagged
Lyophilized from a 0.2?m filtered concentrated (1.0 mg/ml) solution in 20mM NaAc, pH5.5, 4% mannitol.
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
VYLSECKTGN GKNYRGTMSK TKNGITCQKW SSTSPHRPRF SPATHPSEGL EENYCRNPDN DPQGPWCYTT DPEKRYDYCD ILECEEECMH CSGENYDGKI SKTMSGLECQ AWDSQSPHAH GYIPSKFPNK NLKKNYCRNP DRELRPWCFT TDPNKRWELC DIPRCTTPPP SSGPTYQCLKGTGENYRGNV AVTVSGHTCQ HWSAQTPHTH NRTPENFPCK NLDENYCRNP DGKRAPWCHT TNSQVRWEYC KIPSCDSSP
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote