missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ADAT2 Protein

A cDNA sequence encoding the ADAT2 was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP10964-1mg

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965708

  • 7525.00 CHF / 1 Milligramm
Verfügbar ab: 29-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Spezifikation

Spezifikation

134637
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MEAKAAPKPA ASGACSVSAE ETEKWMEEAM HMAKEALENT EVPVGCLMVY NNEVVGKGRN EVNQTKNATR HAEMVAIDQV LDWCRQSGKS PSEVFEHTVL YVTVEPCIMC AAALRLMKIP LVVYGCQNER FGGCGSVLNI ASADLPNTGR PFQCIPGYRA EEAVEMLKTF YKQENPNAPK SKVRKKECQK S
Greater than 90% as determined by Analysis by SDS-PAGE.
ADAT2 Protein
Research Use Only
ADAT2
Human
His
The ADAT2 (0.5 mg/ml) protein contains 20mM Tris-HCl buffer pH 8.0, 2mM DTT, 50mM NaCl and 10% glycerol.
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote