missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 5-hydroxytryptamine receptor 3D Protein
A DNA sequence encoding the Homo sapiens(Human) 5-hydroxytryptamine receptor 3D, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10003-iv-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001138615.1 | |
5-hydroxytryptamine receptor 3D Protein | |
Research Use Only | |
HTR3D | |
Recombinant Protein | |
E. coli | |
VAIRRRCRPS PYVVNFLVPS GILIAIDALS FYLPLESGNC APFKMTVLLG YSVFLLMMND LLPATSTSSH ASLVAPLALM QTPLPAGVYF ALCLSLMVGS LLETIFITHL LHVATTQPLP LPRWLHSLLL HCTGQGRCCP TAPQKGNKGP GLTPTHLPGV KEPEVSAGQM PGPGEAELTG GSEWTRAQRE HEAQKQHSVE LWVQFSHAMD ALLFRLYLLF MASSIITVIC LWNT | |
Greater than 85% as determined by SDS-PAGE. |
200909 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |