missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HSV-8 M Protein

A cDNA sequence encoding the HSV-8 M was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12349-100ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910139

  • 213.00 CHF / 100 Mikrogramm


Verfügbar ab
12-05-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

HSV HSV-8 M Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
GST
HSV-8 Mosaic protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
100mM NaCl, 0.1% SDS and 50% glycerol.
Videos
Dokumentation