missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HSV HSV-2 gD Protein

A cDNA sequence encoding the HSV-2 gD was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP12344-500ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15910129

  • 830.00 CHF / 500 Mikrogramm


Verfügbar ab
27-03-2020
Zum Warenkorb hinzufügen

Spezifikation

Spezifikation

HSV HSV-2 gD Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HSV
His
50% glycerol, 1.5 M Urea, 0.1% SDS and 100 mM NaCl.
Protein is >95% pure as determined by SDS-PAGE
Research Use Only
Recombinant Protein
E. coli
MANUAL
Videos
Dokumentation