missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-1 gG Protein
A cDNA sequence encoding the HSV-1 gG was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12341-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
HSV HSV-1 gG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/ml) 25mM Tris-HCl pH 7.2, 1mM EDTA, and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
GST | |
HSV-1 gG protein is >95% pure as determined by 10% PAGE (coomassie staining). |