missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Horse Erythropoietin / EPO Protein
A DNA sequence encoding the Equus caballus (Horse) Erythropoietin / EPO, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP5988-ec-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001075294.1 | |
Erythropoietin / EPO Protein | |
Research Use Only | |
EPO | |
Recombinant Protein | |
E. coli | |
APPRLICDSR VLERYILEAR EAENVTMGCA EGCSFGENVT VPDTKVNFYS WKRMEVEQQA VEVWQGLALL SEAILQGQAL LANSSQPSET LRLHVDKAVS SLRSLTSLLR ALGAQKEAIS PPDAASAAPL RTFAVDTLCK LFRIYSNFLR GKLKLYTGEA CRRGDR | |
Greater than 90% as determined by SDS-PAGE. |
100033849 | |
100 μg | |
Not determined. | |
Full Length | |
Horse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |