Learn More
enQuireBio™ Recombinant HIV Type-O Envelope Protein
Recombinant Protein
Marke: enQuireBio™ QP12237-100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
HIV Type-O Envelope Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1 mg/ml in water. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |