missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 Protease Protein
A cDNA sequence encoding the HIV-1 Protease was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12260-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
HIV-1 Protease Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
PQITLWQRPL VTIKIGGQLK EALLDTGADD TVLEEMNLPG RWKPKMIGGI GGFIKVRQYD QILIEICGHK AIGTVLVGPT PVNIIGRNLL TQIGCTLNF | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
The HIV-1 Protease filtered (0.4microm) solution is formulated in 20mM Tris, 20mM MES, 200mM NaCl, 1mM EDTA, 10% (v/v) glycerol and 0.05% 2-mercaptoethanol, pH 6.5. |