missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-3 Protein
A cDNA sequence encoding the HCV NS5 Genotype-3 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12192-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
Hepatitis C HCV NS5 Genotype-3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 1.5M urea and 50% glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST | |
HCV NS5 Genotype-3 protein is >95% pure as determined by 10% PAGE (coomassie staining). |