missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Protein
A cDNA sequence encoding the HCV Core was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12138-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Hepatitis C HCV Core Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 1mM EDTA, 1.5M urea and 50%glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV Core protein is >95% pure as determined by 10% PAGE (coomassie staining). |