missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli PPA Protein
A cDNA sequence encoding the PPA was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13112-1mg
Additional Details : Gewicht : 0.01000kg
Spezifikation
PPA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSLLNVPAGK DLPEDIYVVI EIPANADPIK YEIDKESGAL FVDRFMSTAM FYPCNYGYIN HTLSLDGDPV DVLVPTPYPL QPGSVIRCRP VGVLKMTDEA GEDAKLVAVP HSKLSKEYDH IKDVNDLPEL LKAQIAHFFE HYKDLEKGKW VKVEGWENAE AAKAEIVASF ERAKNK | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
PPA protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl. |