missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus, ST3 Dengue NS1 Protein
A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10496-500ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
Dengue Virus, ST3 Dengue NS1 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |
500 μg | |
Untested | |
Dengue Virus, ST3 | |
His | |
>85% as determined by SDS-PAGE. |