missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus, ST3 Dengue NS1 Protein
A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.
870.00 CHF - 1735.00 CHF
Spezifikation
Name | Dengue Virus, ST3 Dengue NS1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Dengue Virus, ST3 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15984048
|
enQuireBio™
QP104961-500UG |
500 μg |
870.00 CHF
500 Mikrogramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15974048
|
enQuireBio™
QP104961-1MG |
1 mg |
1735.00 CHF
1 Milligramm |
Verfügbar ab: 17-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Dengue Virus, ST3 Dengue NS1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus, ST3 | |
His | |
Protein is >90% pure as determined by 12% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline and 50mM arginine. |