Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 1 VLP Protein
Recombinant Protein
Marke:  enQuireBio™ QP10476-500ug
Additional Details : Gewicht : 0.01000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15983988
  • 4010.00 CHF / 500 Mikrogramm
Verfügbar ab
02-11-2020
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Spezifikation
Spezifikation
Serotype 1 VLP Protein
500 μg
Untested
Dengue virus (Strain: Puerto Rico/US/BID-V1053/1998)
Native
>95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
Human
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.