Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 1 VLP Protein
Recombinant Protein
Marke: enQuireBio™ QP10476-100ug
Additional Details : Gewicht : 0.01000kg
Beschreibung
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.Spezifikation
Serotype 1 VLP Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4. |
100 μg | |
Untested | |
Dengue virus (Strain: Puerto Rico/US/BID-V1053/1998) | |
Native | |
>95% as determined by SDS-PAGE. |