missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chlamydia HSP70 Protein

A cDNA sequence encoding the Chlamydia HSP70 (Amino Acids 549-660) was constructed and used to recombinantly synthesize the protein.

Marke:  enQuireBio™ QP14020-EC-500ug

Additional Details : Gewicht : 0.01000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15965429

Menge Preis
1 852.02 CHF / 500 Mikrogramm
5 783.52 CHF / 500 Mikrogramm
Voraussichtliches Versanddatum: 28-10-2021

Zum Warenkorb hinzufügen


Beschreibung

Beschreibung

Spezifikation

Spezifikation

HSP70 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Chlamydia
Untagged
Chlamydia HSP70 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC.
Research Use Only
Recombinant Protein
E. coli
50mM Tris-HCl, pH 8.0, 8M Urea, 60mM NaCl and 50% glycerol.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate