missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine HGFR/c-MET Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 4140-ME-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Canine HGF R/c-MET Protein is derived from NS0. The Recombinant Canine HGF R/c-MET Protein has been validated for the following applications: Binding Activity.
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
HGFR/c-MET | |
Mouse myeloma cell line,NS0-derived canine HGF R/c-MET protein Glu25-Arg308 ( alpha chain) & Ser309-Leu935 ( beta chain),with a C-terminal 6-His tag | |
AUTS9, c-Met, cMET, EC 2.7.10, EC 2.7.10.1, hepatocyte growth factor receptor, HGF R, HGF receptor, HGF/SF receptor, HGFR, MET, Met (c-Met), met proto-oncogene (hepatocyte growth factor receptor), met proto-oncogene tyrosine kinase, oncogene MET, Proto-oncogene c-Met, RCCP2, Scatter factor receptor, SF receptor, Tyrosine-protein kinase Met | |
Recombinant |
32.6 kDa ( alpha chain) and 70.1 kDa (beta chain) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
R&D Systems™ Recombinant Canine HGFR/c-MET Protein > 50ug; Carrier Free
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur