missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine FGF 2 Protein
A cDNA sequence encoding the FGF 2 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP10599-1mg
Additional Details : Gewicht : 0.01000kg
Especificaciones
281161 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The recommended concentration in responsive cells is 0.1 to 2ng/ml. | |
Bovine | |
Untagged | |
The FGF-basic Bovine was lyophilized from a concentrated sterile solution containing 50mM Na2HPO4, pH 7.5 and 0.5% HSA. |
FGF 2 Protein | |
Research Use Only | |
FGF2 | |
Recombinant Protein | |
Bovine | |
MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGPKTG PGQKAILFLP MSAKS | |
Greater than 95.0% as determined by SDS-PAGE. |