Learn More
Heidolph™ Reax Top with Small Pick-Up Attachment
Artikelnummer. 17383450 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17383450 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17383450 Lieferant Heidolph™ Lieferanten-Nr. 5411000002
Verfügbar ab: 05-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Reax Top is particularly suitable for simple mixing tasks.
  • 5 mm shaking orbit
  • Particularly suitable for simple mixing tasks
  • In short-term operation mode, the shaking motion is triggered by pressure on the test tube tray
  • This mode is suitable for mixing up solutions, for example
  • Persistent cell pellets can be dissolved gently but powerfully in continuous operation
  • With a speed of up to 2500 rpm and a shaking orbit of 5 mm, reliable and uniform distribution of solids, and homogenization of highly viscous media are guaranteed
TRUSTED_SUSTAINABILITY
Spezifikation
Spannung 230 V
Steckertyp CH-Stecker