Learn More
Heidolph™ Reax Control with Small Pick-Up Attachment
Artikelnummer. 17393450 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17393450 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17393450 Lieferant Heidolph™ Lieferanten-Nr. 5411100002
Verfügbar ab: 09-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Reax Control is a precise model with speed scaling.
  • All the outstanding features of the Reax Top can be found in this model: a rotational speed of up to 2500 rpm and a 5 mm shaking orbit
  • The electronic speed control and the scaling of the speed display allow precise settings
  • Applications that require a determined speed are thus shaken more gently
  • Shear force-sensitive organisms remain particularly protected by the constant speed in the low range
TRUSTED_SUSTAINABILITY
Spezifikation
Spannung 230 V
Steckertyp CH-Stecker