Learn More
Heidolph™ Reax 20/12
Artikelnummer. 17323460 Alle ansehen Heidolph Instruments Produkte
Ansicht ändern
Click to view available options
Steckertyp:
CH-Stecker
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Steckertyp Packungsgröße
17323460 CH-Stecker 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17323460 Lieferant Heidolph™ Lieferanten-Nr. 5412001208
Verfügbar ab: 26-11-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Resilient shaker that can hold up to 12 bottles.
  • The Reax 20 is an ideal shaker for processing and storing samples that tend to sediment. The fields of application range from environmental technology to paint processing and quality assurance in the materials industry.
  • The maximum permissible weight is 30 kg, so that samples with a high solid content can also be properly mixed. Since this device sets great forces in motion, it is equipped with an emergency stop switch for your safety.
  • This overhead shaker can be ordered with a capacity of 4, 8 or 12 bottles. Also suitable for mixing cylinders or wide-neck bottles.
TRUSTED_SUSTAINABILITY
Spezifikation
Spannung 230 V
Steckertyp CH-Stecker