Learn More
Invitrogen™ ReadyLabel™ Antibody Labeling Kits
Artikelnummer. 18041663
Click to view available options
Menge:
5 x 100 μg of IgG
5 x 20 μg
5 x 20 μg of IgG
Marker oder Farbstoff:
Alexa Fluor 594
Alexa Fluor Plus 555
Alexa Fluor Plus 750
Alexa Fluor 488
Alexa Fluor 514
Alexa Fluor 647
Alexa Fluor 700
Biotin
Keine
Packungsgröße:
5 Reaktionen
5 Stück
Artikelnummer. 18041663
Marke: Invitrogen™ R10702
om dit product te kopen Registreer vandaag om een webaccount aan te maken
ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution.
ReadyLabel Antibody Labeling Kits enable fast, convenient labeling of off-the-shelf antibodies and cell culture supernatants without prior purification of the antibody solution. Each kit contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step.
Each ReadyLabel Antibody Labeling Kit, 100 μg contains everything needed to perform 5 labeling reactions and will produce purified antibody conjugates, without the purification step. The kit can be used to label antibodies in solutions containing BSA or other stabilizing proteins and antibodies in a primary amine-containing buffer such as Tris or glycine, or even cell culture supernatants, without prior purification of the antibody-containing solution. A reactive dye moiety efficiently binds to primary amines of antibodies to form stable, covalently bound antibody-dye conjugates.
Alexa Fluor dyes produce antibody conjugates that are typically brighter with better photostability and lower pH sensitivity relative to traditional dyes (e.g., FITC). Unlike other dyes, the fluorescence of Alexa Fluor conjugates are insensitive to pH between 4 and 10. Alexa Fluor-labeled antibodies can be used for applications including microscopy, flow cytometry, western blotting, ELISA, and indirect FISH.
ReadyLabel Antibody Labeling Kits offer the following benefits:
  • Convenience—Labeling takes one hour, with less than 20 minutes of hands-on time
  • Versatility—Labelling can be performed on IgG antibodies, regardless of species, isotype, or storage buffer
  • Customization—ReadyLabel Flex Antibody Labeling Kits are designed for use with a wide range of amine-reactive labels including Alexa Fluor dyes
TRUSTED_SUSTAINABILITY
Spezifikation
Zur Verwendung mit (Anwendung) Antibody Production and Labeling
Produkttyp Antikörpermarkierungskit
Menge 5 x 100 μg of IgG
Zur Verwendung mit (Geräte) Durchflusszytometer
Produktlinie ReadyLabel™ Flex
Chemische Reaktivität Amine
Löslichkeit DMSO (Dimethylsulfoxid)
Marker oder Farbstoff Keine
Farbe Farblos
Nur für Forschungszwecke.