missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ready-Made Terrific Broth Powder

Provides greater yields of plasmid DNA and recombinant proteins than LB media by allowing for greater cell density

Marke:  Thermo Scientific™ J75856.A1

Additional Details : Gewicht : 1.00000kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15815378

  • 138.67 CHF / 1 Kilogramm


Geschätzter Ausliefertermin
von Anbieter 18-03-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

  • Off-white free flowing powder
  • pH(5.08%, H2O): 7.2 ± 0.2
Contents
  • Casein Peptone: 12g/L
  • Yeast Extract: 24g/L
  • Potassium Phosphate Dibasic: 12.5g/L
  • Potassium Phosphate Monobasic: 2.3g/L
Spezifikation

Spezifikation

Off-white
Plastic Bottle
Terrific Broth
Free Flowing Powder
1kg
Provides greater yields of plasmid DNA and recombinant proteins
Videos