missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RDM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5110906
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: SPVKVRLGTR HKAVQHQALA LNSSKCQELA NYYFGFNGCS KRIIKLQELS DLEERENEDS MVPLPKQSLK FFCALEVVLP SCDCRSPGIG LVE Highest antigen sequence indentity to the following orthologs: Rat - 70%, Mouse - 71%.
This gene encodes a protein involved in the cellular response to cisplatin, a drug commonly used in chemotherapy. The protein encoded by this gene contains two motifs: a motif found in RAD52, a protein that functions in DNA double-strand breaks and homologous recombination, and an RNA recognition motif (RRM) that is not found in RAD52. The RAD52 motif region in RAD52 is important for protein function and may be involved in DNA binding or oligomerization. Alternatively spliced transcript variants encoding different isoforms have been reported.
Spezifikation
| RDM1 | |
| Polyclonal | |
| Unconjugated | |
| RDM1 | |
| 2410008M22Rik; AW212028; DeltaN-RDM1zeta; RAD52 homolog B; RAD52 motif 1; RAD52 motif containing 1; RAD52 motif-containing protein 1; RAD52B; RAD52B homolog; Rdm1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 201299 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NG50 | |
| RDM1 | |
| Recombinant Protein corresonding to Human RDM1. Recombinant protein control fragment (Product #RP-93528). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur