missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11774-1-AP-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This protein belongs to the lipocalin family and is the specific carrier for retinol (vitamin A alcohol) in the blood. It delivers retinol from the liver stores to the peripheral tissues. In plasma, the RBP-retinol complex interacts with transthyretin which prevents its loss by filtration through the kidney glomeruli. A deficiency of vitamin A blocks secretion of the binding protein posttranslationally and results in defective delivery and supply to the epidermal cells.Spezifikation
RBP4 | |
Polyclonal | |
Unconjugated | |
RBP4 | |
Plasma retinol binding protein, PRBP, RBP, RBP4, Retinol binding protein 4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19662, 25703, 5950 | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02753, P04916, Q00724 | |
RBP4 | |
RBP4 Fusion Protein Ag2448 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Liquid |