missing translation for 'onlineSavingsMsg'
Få mere at vide

RBBP9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Artikelnummer. p-7203730 Shop alle Proteintech produkter
Skift visning
Klik for at se tilgængelige muligheder
Menge:
20 μL
150 μL
Pakningsstørrelse:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16819823 20 μL 20 Mikroliter
16809823 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16819823 Leverandør Proteintech Leverandørnr. 122302AP20UL

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.

Tekniske data

Antigen RBBP9
Anwendungen Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry
Klassifikation Polyclonal
Konzentration 0.2 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gen RBBP9
Gen-Zugriffsnummer O75884, O88350, O88851
Gen-Alias B5T overexpressed gene protein, BOG, Protein BOG, Putative hydrolase RBBP9, RBBP 10, RBBP 9, RBBP10, RBBP9
Gensymbole Rbbp9
Wirtsspezies Rabbit
Immunogen RBBP9 Fusion Protein Ag2868
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 10741, 26450, 29459
Zielspezies Human, Rat, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.