missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Rat LAG3 / CD223 Protein, His Tag
365.69 CHF - 2774.20 CHF
Spezifikation
Zugriffsnummer | NP_997678 |
---|---|
Molekulargewicht | 47.6 kDa |
Quelle | Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession # NP_997678). |
Immunogen | Gly 24 - Leu 442 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15807227
|
ACROBiosystems
LA3-R52H5-100UG |
100 μg |
365.69 CHF
100 Mikrogramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817227
|
ACROBiosystems
LA3-R52H5-1MG |
1 mg |
2774.20 CHF
1 Milligramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Lymphocyte activation gene 3 protein (LAG3) is also known as CD antigen CD223 and protein FDC, which belongs to immunoglobulin (Ig) superfamily and contains 4 extracellular Ig-like domains. The LAG3 gene contains 8 exons. The sequence data, exon/intron organization, and chromosomal localization all indicate a close relationship of LAG3 to CD4. LAG3 /CD223 involved in lymphocyte activation. LAG3 /CD223 binds to HLA class-II antigens.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.6 kDa. The protein migrates as 55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_997678 | |
Rat LAG3, His Tag (LA3-R52H5) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 442 (Accession # NP_997678). | |
-20°C | |
LAG-3 | |
Rat | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
47.6 kDa | |
Gly 24 - Leu 442 | |
LAG3,CD223,FDC | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |