Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), Super Bright™ 780, eBioscience™, Invitrogen™
Rat Isotype Control
156.00 CHF - 365.00 CHF
Spezifikation
Antigen | Rat IgG2b kappa |
---|---|
Klon | eB149/10H5 |
Konjugat | Super Bright 780 |
Form | Liquid |
Wirtsspezies | Rat |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15924174
|
Affymetrix eBioscience
78-4031-82 |
100 μg |
365.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
15914174
|
Affymetrix eBioscience
78-4031-80 |
25 μg |
Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11. |
||||||
Beschreibung
Description: The rat IgG2b monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: This eB149/10H5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This eB149/10H5 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells and mouse splenocytes. Use at the same concentration as the experimental antibody. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405nm) and emits at 780nm. We recommend using a 780/60 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (cat. SB-4400) to minimize any non-specific polymer interactions. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100μL of cell sample + 100μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 405nm; Emission: 780nm; Laser: Violet Laser
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Rat IgG2b kappa | |
Super Bright 780 | |
Rat | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
eB149/10H5 | |
Liquid | |
RUO | |
IgG2b κ | |
Affinity chromatography |