missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 450, eBioscience™, Invitrogen™
Rat Isotype Control
111.00 CHF - 254.00 CHF
Spezifikation
Antigen | Rat IgG2b kappa |
---|---|
Klon | eB149/10H5 |
Konzentration | 0.2 mg/mL |
Anwendungen | Control, Flow Cytometry |
Konjugat | eFluor 450 |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15301770
|
Affymetrix eBioscience
48-4031-80 |
25 μg |
111.00 CHF
25 Mikrogramm |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15311770
|
Affymetrix eBioscience
48-4031-82 |
100 μg |
254.00 CHF
100 Mikrogramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Description: The rat IgG2b, kappa monoclonal antibody is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG2b Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Rat IgG2b Isotype Control has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2b kappa Control antibody specifically detects Rat IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Spezifikation
Rat IgG2b kappa | |
0.2 mg/mL | |
eFluor 450 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eB149/10H5 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2b κ | |
Affinity chromatography |
For Research Use Only.